The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000208.3 | NP_000199.2 | 2 - 3 | 1062 | 75 |
NM_001079817.2 | NP_001073285.1 | 2 - 3 | 1062 | 75 | |
XM_011527988.2 | XP_011526290.2 | 2 - 3 | 1165 | 75 | |
XM_011527989.2 | XP_011526291.2 | 2 - 3 | 1164 | 75 | |
GenBank mRNA | BC047591.1 | - | 2 - 3 | 626 | 75 |
BC117172.1 | - | 2 - 3 | 684 | 75 | |
M10051.1 | - | 2 - 3 | 789 | 75 | |
X02160.1 | - | 2 - 3 | 699 | 75 |