The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001066.2 | NP_001057.1 | 2 - 3 | 271 | 82 |
XM_011542060.2 | XP_011540362.1 | 2 - 3 | 326 | 82 | |
XM_011542063.2 | XP_011540365.1 | 2 - 3 | 326 | 82 | |
XM_017002211.1 | XP_016857700.1 | 2 - 3 | 326 | 82 | |
XM_017002212.1 | XP_016857701.1 | 3 - 4 | 1110 | 82 | |
XM_017002213.1 | XP_016857702.1 | 4 - 5 | 1149 | 82 | |
GenBank mRNA | AY148473.1 | - | 2 - 3 | 188 | 82 |
BC052977.1 | - | 2 - 3 | 269 | 82 | |
BT019927.1 | - | 2 - 3 | 182 | 82 | |
EU826615.1 | - | 2 - 3 | 182 | 82 | |
M32315.1 | - | 2 - 3 | 271 | 82 | |
M55994.1 | - | 2 - 3 | 274 | 82 | |
S63368.1 | - | 1 - 2 | 74 | 82 |