Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNFRSF1B
Entrez Gene ID
7133
Gene Name
TNF receptor superfamily member 1B
Gene Aliases
CD120b, TBPII, TNF-R-II, TNF-R75, TNFBR, TNFR1B, TNFR2, TNFR80, p75, p75TNFR
Chromosome Location
Chr.1: 12166948 - 12209222 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001066.2 NP_001057.1 3 - 4 395 70
XM_011542060.2 XP_011540362.1 3 - 4 450 70
XM_011542063.2 XP_011540365.1 3 - 4 450 70
XM_017002211.1 XP_016857700.1 3 - 4 450 70
XM_017002212.1 XP_016857701.1 4 - 5 1234 70
XM_017002213.1 XP_016857702.1 5 - 6 1273 70
XM_017002214.1 XP_016857703.1 2 - 3 350 70
XM_017002215.1 XP_016857704.1 2 - 3 350 70
GenBank mRNA BC011844.2 - 2 - 3 295 70
BC042167.1 - 2 - 3 295 70
BC052977.1 - 3 - 4 393 70
BT019927.1 - 3 - 4 306 70
EU826615.1 - 3 - 4 306 70
M32315.1 - 3 - 4 395 70
M55994.1 - 3 - 4 398 70
S63368.1 - 2 - 3 198 70

View Full Product Details