Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM208A
Entrez Gene ID
23272
Gene Name
family with sequence similarity 208 member A
Gene Aliases
C3orf63, RAP140, se89-1
Chromosome Location
Chr.3: 56620132 - 56683242 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112736.1 NP_001106207.1 12 - 13 1547 130
NM_015224.3 NP_056039.2 5 - 6 898 130
XM_005264999.1 XP_005265056.1 12 - 13 1682 130
XM_006713077.2 XP_006713140.1 12 - 13 1682 130
XM_006713078.2 XP_006713141.1 6 - 7 536 130
XM_011533552.2 XP_011531854.1 12 - 13 1682 130
XM_011533553.2 XP_011531855.1 11 - 12 1103 130
XM_017006030.1 XP_016861519.1 12 - 13 1457 130
XM_017006031.1 XP_016861520.1 12 - 13 1682 130
XM_017006032.1 XP_016861521.1 11 - 12 1103 130
GenBank mRNA AF180425.2 - 5 - 6 1182 130
AK094486.1 - 12 - 13 1677 130
AK307599.1 - 4 - 5 360 130
BC029354.1 - 5 - 6 898 130
BC070096.1 - 12 - 13 1545 130
BC129987.1 - 4 - 5 323 130
BX648955.1 - 11 - 12 1029 130

View Full Product Details