The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001112736.1 | NP_001106207.1 | 1 - 2 | 434 | 65 |
XM_005264999.1 | XP_005265056.1 | 1 - 2 | 569 | 65 | |
XM_006713077.2 | XP_006713140.1 | 1 - 2 | 569 | 65 | |
XM_011533552.2 | XP_011531854.1 | 1 - 2 | 569 | 65 | |
XM_011533553.2 | XP_011531855.1 | 1 - 2 | 115 | 65 | |
XM_017006030.1 | XP_016861519.1 | 1 - 2 | 344 | 65 | |
XM_017006031.1 | XP_016861520.1 | 1 - 2 | 569 | 65 | |
XM_017006032.1 | XP_016861521.1 | 1 - 2 | 115 | 65 | |
GenBank mRNA | AK094486.1 | - | 1 - 2 | 564 | 65 |
BC070096.1 | - | 1 - 2 | 432 | 65 |