The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001112736.1 | NP_001106207.1 | 22 - 23 | 4421 | 100 |
NM_015224.3 | NP_056039.2 | 15 - 16 | 3649 | 100 | |
XM_005264999.1 | XP_005265056.1 | 22 - 23 | 4556 | 100 | |
XM_006713077.2 | XP_006713140.1 | 22 - 23 | 4433 | 100 | |
XM_006713078.2 | XP_006713141.1 | 16 - 17 | 3410 | 100 | |
XM_011533552.2 | XP_011531854.1 | 21 - 22 | 4373 | 100 | |
XM_011533553.2 | XP_011531855.1 | 21 - 22 | 3977 | 100 | |
XM_017006030.1 | XP_016861519.1 | 22 - 23 | 4208 | 100 | |
XM_017006031.1 | XP_016861520.1 | 21 - 22 | 4373 | 100 | |
XM_017006032.1 | XP_016861521.1 | 21 - 22 | 3854 | 100 | |
GenBank mRNA | AB029028.1 | - | 5 - 6 | 1502 | 100 |
AF180425.2 | - | 15 - 16 | 3933 | 100 | |
AF273053.1 | - | 8 - 9 | 2573 | 100 | |
AK307599.1 | - | 14 - 15 | 3111 | 100 | |
BC070096.1 | - | 21 - 22 | 4237 | 100 | |
BC129987.1 | - | 14 - 15 | 3197 | 100 | |
BX648955.1 | - | 21 - 22 | 3903 | 100 |