Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM208A
Entrez Gene ID
23272
Gene Name
family with sequence similarity 208 member A
Gene Aliases
C3orf63, RAP140, se89-1
Chromosome Location
Chr.3: 56620132 - 56683242 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015224.3 NP_056039.2 16 - 17 3811 90
XM_005264999.1 XP_005265056.1 23 - 24 4718 90
XM_006713077.2 XP_006713140.1 23 - 24 4595 90
XM_006713078.2 XP_006713141.1 17 - 18 3572 90
XM_011533552.2 XP_011531854.1 22 - 23 4535 90
XM_011533553.2 XP_011531855.1 22 - 23 4139 90
GenBank mRNA AB029028.1 - 6 - 7 1664 90
AF180425.2 - 16 - 17 4095 90
AF273053.1 - 9 - 10 2735 90
AK307599.1 - 15 - 16 3273 90
BC070096.1 - 22 - 23 4399 90

View Full Product Details