The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030907.3 | NP_112169.2 | 3 - 4 | 814 | 85 |
XM_011542126.2 | XP_011540428.1 | 3 - 4 | 1095 | 85 | |
GenBank mRNA | AL559024.3 | - | 3 - 4 | 744 | 85 |
BC002946.2 | - | 3 - 4 | 601 | 85 | |
BC008702.1 | - | 3 - 4 | 601 | 85 |