Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTRC
Entrez Gene ID
11330
Gene Name
chymotrypsin C
Gene Aliases
CLCR, ELA4
Chromosome Location
Chr.1: 15438442 - 15449247 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007272.2 NP_009203.2 4 - 5 382 75
XM_011540550.1 XP_011538852.1 4 - 5 382 75
GenBank mRNA AK289447.1 - 4 - 5 381 75
BC015118.1 - 4 - 5 388 75
BC059779.1 - 4 - 5 361 75
CR749296.1 - 2 - 3 192 75
S82198.1 - 4 - 5 359 75
Y13697.1 - 3 - 4 311 75

View Full Product Details