The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007272.2 | NP_009203.2 | 6 - 7 | 663 | 136 |
GenBank mRNA | AK289447.1 | - | 6 - 7 | 662 | 136 |
BC015118.1 | - | 6 - 7 | 669 | 136 | |
S82198.1 | - | 6 - 7 | 640 | 136 | |
Y13697.1 | - | 5 - 6 | 592 | 136 |