Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRIQ3
Entrez Gene ID
127255
Gene Name
leucine rich repeats and IQ motif containing 3
Gene Aliases
LRRC44
Chromosome Location
Chr.1: 74026015 - 74198201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105659.1 NP_001099129.1 5 - 6 1049 140
NM_001322315.1 NP_001309244.1 5 - 6 1049 140
XM_011540658.1 XP_011538960.1 4 - 5 739 140
XM_011540659.1 XP_011538961.1 3 - 4 253 140

View Full Product Details