The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000186.3 | NP_000177.2 | 10 - 11 | 1758 | 110 |
GenBank mRNA | AK226113.1 | - | 5 - 6 | 3544 | 110 |
AK291395.1 | - | 10 - 11 | 1597 | 110 | |
BC142699.1 | - | 10 - 11 | 1570 | 110 | |
BP324193.1 | - | 2 - 3 | 328 | 110 | |
Y00716.1 | - | 10 - 11 | 1591 | 110 |