The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000186.3 | NP_000177.2 | 17 - 18 | 3020 | 92 |
GenBank mRNA | AK226113.1 | - | 12 - 13 | 4806 | 92 |
AK291395.1 | - | 17 - 18 | 2859 | 92 | |
BC142699.1 | - | 17 - 18 | 2832 | 92 | |
M17517.1 | - | 6 - 7 | 1059 | 92 | |
Y00716.1 | - | 17 - 18 | 2853 | 92 |