The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293312.1 | NP_001280241.1 | 3 - 4 | 479 | 68 |
NM_001349.3 | NP_001340.2 | 4 - 5 | 537 | 68 | |
XM_017003489.1 | XP_016858978.1 | 4 - 5 | 508 | 68 | |
GenBank mRNA | AK129521.1 | - | 4 - 5 | 479 | 68 |
AK222476.1 | - | 4 - 5 | 367 | 68 | |
AK290607.1 | - | 4 - 5 | 394 | 68 | |
AY762100.1 | - | 4 - 5 | 356 | 68 | |
BC000629.2 | - | 4 - 5 | 370 | 68 | |
BC107749.1 | - | 4 - 5 | 350 | 68 | |
BI753718.1 | - | 4 - 5 | 509 | 68 | |
BT006710.1 | - | 4 - 5 | 321 | 68 | |
J05032.1 | - | 4 - 5 | 411 | 68 |