The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011552.3 | NP_001011552.2 | 2 - 3 | 1175 | 109 |
XM_011511158.1 | XP_011509460.1 | 2 - 3 | 1196 | 109 | |
GenBank mRNA | CR627411.1 | - | 2 - 3 | 1176 | 109 |