The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011552.3 | NP_001011552.2 | 8 - 9 | 2177 | 102 |
XM_011511158.1 | XP_011509460.1 | 8 - 9 | 2111 | 102 | |
GenBank mRNA | CR627411.1 | - | 8 - 9 | 2178 | 102 |