Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGEF26
Entrez Gene ID
26084
Gene Name
Rho guanine nucleotide exchange factor 26
Gene Aliases
CSGEF, HMFN1864, SGEF
Chromosome Location
Chr.3: 154121003 - 154257827 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001251962.1 NP_001238891.1 12 - 13 2585 76
NM_001251963.1 NP_001238892.1 12 - 13 2512 76
NM_015595.3 NP_056410.3 12 - 13 2512 76
XM_011512672.1 XP_011510974.1 11 - 12 2422 76
XM_017006160.1 XP_016861649.1 11 - 12 1254 76
GenBank mRNA AF415175.1 - 12 - 13 2467 76
AF415176.1 - 2 - 3 437 76
AK022655.1 - 6 - 7 823 76
AK022884.1 - 9 - 10 1099 76
AK123040.1 - 12 - 13 2456 76
AL117429.1 - 2 - 3 323 76
AY552599.1 - 11 - 12 2301 76
BC016628.1 - 6 - 7 731 76
BC078655.1 - 12 - 13 2453 76

View Full Product Details