The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003246.3 | NP_003237.2 | 9 - 10 | 1652 | 67 |
GenBank mRNA | AB209912.1 | - | 9 - 10 | 1638 | 67 |
AK291639.1 | - | 9 - 10 | 1652 | 67 | |
AK304754.1 | - | 9 - 10 | 1398 | 67 | |
BC136469.1 | - | 9 - 10 | 1626 | 67 | |
BC136470.1 | - | 9 - 10 | 1626 | 67 | |
X04665.1 | - | 9 - 10 | 1548 | 67 | |
X14787.1 | - | 9 - 10 | 1584 | 67 |