Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ODF2
Entrez Gene ID
4957
Gene Name
outer dense fiber of sperm tails 2
Gene Aliases
CT134, ODF2/1, ODF2/2, ODF84
Chromosome Location
Chr.9: 128455155 - 128501292 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242352.1 NP_001229281.1 20 - 21 2669 77
NM_001242353.1 NP_001229282.1 20 - 21 2755 77
NM_002540.4 NP_002531.3 22 - 23 2957 77
NM_153433.1 NP_702911.1 20 - 21 2682 77
NM_153435.1 NP_702913.1 20 - 21 2575 77
XM_005252010.2 XP_005252067.1 19 - 20 2623 77
XM_005252011.2 XP_005252068.1 19 - 20 2563 77
XM_005252012.3 XP_005252069.1 20 - 21 2813 77
XM_005252013.3 XP_005252070.1 20 - 21 2760 77
XM_005252014.3 XP_005252071.1 20 - 21 2809 77
XM_005252017.4 XP_005252074.1 20 - 21 2714 77
XM_005252018.2 XP_005252075.1 21 - 22 2919 77
XM_005252019.3 XP_005252076.1 21 - 22 2938 77
XM_005252022.2 XP_005252079.1 20 - 21 2737 77
XM_005252023.2 XP_005252080.1 20 - 21 2708 77
XM_005252024.3 XP_005252081.1 20 - 21 2727 77
XM_005252026.4 XP_005252083.1 20 - 21 2656 77
XM_005252029.2 XP_005252086.1 20 - 21 2581 77
XM_006717126.3 XP_006717189.1 20 - 21 2657 77
XM_011518722.2 XP_011517024.1 20 - 21 2737 77
XM_011518723.2 XP_011517025.1 20 - 21 2680 77
XM_011518724.2 XP_011517026.1 21 - 22 2901 77
XM_011518725.2 XP_011517027.1 21 - 22 2905 77
XM_011518726.2 XP_011517028.1 21 - 22 2923 77
XM_011518728.2 XP_011517030.1 20 - 21 2674 77
XM_011518729.2 XP_011517031.1 20 - 21 2610 77
XM_011518730.2 XP_011517032.1 20 - 21 2581 77
XM_011518731.2 XP_011517033.1 19 - 20 2566 77
XM_011518732.2 XP_011517034.1 20 - 21 2523 77
XM_011518733.2 XP_011517035.1 19 - 20 2509 77
XM_017014764.1 XP_016870253.1 21 - 22 2894 77
XM_017014765.1 XP_016870254.1 21 - 22 2913 77
XM_017014766.1 XP_016870255.1 19 - 20 2467 77
XM_017014767.1 XP_016870256.1 21 - 22 2749 77
XM_017014768.1 XP_016870257.1 20 - 21 2697 77
XM_017014769.1 XP_016870258.1 20 - 21 2726 77
XM_017014770.1 XP_016870259.1 21 - 22 2881 77
XM_017014771.1 XP_016870260.1 21 - 22 2862 77
XM_017014772.1 XP_016870261.1 21 - 22 2782 77
XM_017014773.1 XP_016870262.1 21 - 22 2758 77
XM_017014774.1 XP_016870263.1 20 - 21 2468 77
XM_017014775.1 XP_016870264.1 21 - 22 2725 77
XM_017014776.1 XP_016870265.1 21 - 22 2706 77
XM_017014777.1 XP_016870266.1 20 - 21 2508 77
XM_017014778.1 XP_016870267.1 20 - 21 2611 77
XM_017014779.1 XP_016870268.1 19 - 20 2352 77
XM_017014780.1 XP_016870269.1 20 - 21 2535 77
XM_017014781.1 XP_016870270.1 19 - 20 2290 77
GenBank mRNA AK126816.1 - 3 - 4 727 77
AK299303.1 - 20 - 21 2517 77
AK302684.1 - 20 - 21 2609 77
AY366499.1 - 20 - 21 2452 77
BC010629.1 - 22 - 23 2755 77
BC012785.2 - 3 - 4 332 77
BC091500.1 - 20 - 21 2569 77
DQ444713.1 - 18 - 19 2282 77
DQ444714.1 - 18 - 19 2225 77

View Full Product Details