The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164317.1 | NP_001157789.1 | 2 - 3 | 705 | 62 |
NM_001164318.1 | NP_001157790.1 | 2 - 3 | 705 | 62 | |
NM_001164319.1 | NP_001157791.1 | 2 - 3 | 705 | 62 | |
NM_001457.3 | NP_001448.2 | 2 - 3 | 705 | 62 | |
XM_005264978.1 | XP_005265035.1 | 2 - 3 | 685 | 62 | |
GenBank mRNA | AB191258.1 | - | 2 - 3 | 683 | 62 |
AB371580.1 | - | 2 - 3 | 685 | 62 | |
AB371581.1 | - | 2 - 3 | 683 | 62 | |
AB371582.1 | - | 2 - 3 | 683 | 62 | |
AF042166.1 | - | 2 - 3 | 671 | 62 | |
AF043045.1 | - | 2 - 3 | 705 | 62 |