The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164317.1 | NP_001157789.1 | 33 - 34 | 5685 | 101 |
NM_001164318.1 | NP_001157790.1 | 32 - 33 | 5559 | 101 | |
NM_001164319.1 | NP_001157791.1 | 31 - 32 | 5520 | 101 | |
NM_001457.3 | NP_001448.2 | 32 - 33 | 5592 | 101 | |
XM_005264978.1 | XP_005265035.1 | 32 - 33 | 5593 | 101 | |
GenBank mRNA | AB191258.1 | - | 32 - 33 | 5537 | 101 |
AB209889.1 | - | 13 - 14 | 2461 | 101 | |
AB371580.1 | - | 33 - 34 | 5665 | 101 | |
AB371581.1 | - | 31 - 32 | 5498 | 101 | |
AB371582.1 | - | 32 - 33 | 5570 | 101 | |
AF042166.1 | - | 32 - 33 | 5558 | 101 | |
AF043045.1 | - | 32 - 33 | 5592 | 101 | |
BX641085.1 | - | 31 - 32 | 5117 | 101 | |
CR749793.1 | - | 30 - 31 | 5005 | 101 |