Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DHX36
Entrez Gene ID
170506
Gene Name
DEAH-box helicase 36
Gene Aliases
DDX36, G4R1, MLEL1, RHAU
Chromosome Location
Chr.3: 154275668 - 154324497 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114397.1 NP_001107869.1 2 - 3 444 85
NM_020865.2 NP_065916.2 2 - 3 444 85
GenBank mRNA AF217190.1 - 2 - 3 436 85
AJ577133.1 - 2 - 3 363 85
AJ577134.1 - 2 - 3 363 85
AK303720.1 - - 324 85
AK314435.1 - 2 - 3 444 85
BC036035.1 - 2 - 3 408 85

View Full Product Details