The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300845.1 | NP_001287774.1 | 5 - 6 | 1224 | 108 |
NM_173508.3 | NP_775779.1 | 6 - 7 | 1509 | 108 | |
GenBank mRNA | AK095031.1 | - | 6 - 7 | 1290 | 108 |
BC037878.1 | - | 5 - 6 | 1168 | 108 |