The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193397.1 | NP_001180326.1 | 8 - 9 | 856 | 90 |
NM_001271832.1 | NP_001258761.1 | 7 - 8 | 983 | 90 | |
NM_021131.4 | NP_066954.2 | 9 - 10 | 996 | 90 | |
NM_178000.2 | NP_821067.1 | 8 - 9 | 1070 | 90 | |
NM_178001.2 | NP_821068.1 | 9 - 10 | 1175 | 90 | |
NM_178003.2 | NP_821070.1 | 7 - 8 | 944 | 90 | |
XM_011518834.2 | XP_011517136.1 | 9 - 10 | 995 | 90 | |
XM_011518835.2 | XP_011517137.1 | 9 - 10 | 1153 | 90 | |
XM_011518836.2 | XP_011517138.1 | 8 - 9 | 909 | 90 | |
XM_011518837.2 | XP_011517139.1 | 9 - 10 | 1153 | 90 | |
XM_011518838.2 | XP_011517140.1 | 8 - 9 | 890 | 90 | |
XM_017014888.1 | XP_016870377.1 | 8 - 9 | 1048 | 90 | |
GenBank mRNA | AK222788.1 | - | 8 - 9 | 969 | 90 |
AK297202.1 | - | 7 - 8 | 660 | 90 | |
AK302897.1 | - | 9 - 10 | 1101 | 90 | |
AK307424.1 | - | 8 - 9 | 855 | 90 | |
AU125192.1 | - | 5 - 6 | 550 | 90 | |
BC002545.2 | - | 9 - 10 | 980 | 90 | |
BC011605.2 | - | 9 - 10 | 980 | 90 | |
BQ896712.1 | - | 3 - 4 | 320 | 90 | |
BT020119.1 | - | 8 - 9 | 783 | 90 | |
CD630703.1 | - | 2 - 3 | 90 | ||
X73478.1 | - | 8 - 9 | 972 | 90 |