The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002890.2 | NP_002881.1 | 10 - 11 | 1658 | 152 |
NM_022650.2 | NP_072179.1 | 10 - 11 | 1052 | 152 | |
XM_011543525.1 | XP_011541827.1 | 10 - 11 | 2110 | 152 | |
XM_011543527.2 | XP_011541829.1 | 10 - 11 | 2110 | 152 | |
GenBank mRNA | AB209106.1 | - | 10 - 11 | 1658 | 152 |
AK300263.1 | - | 11 - 12 | 1159 | 152 | |
AK300269.1 | - | 10 - 11 | 985 | 152 | |
AK300406.1 | - | 11 - 12 | 1078 | 152 | |
AK312739.1 | - | 10 - 11 | 1037 | 152 | |
BC033015.2 | - | 10 - 11 | 1646 | 152 | |
BC107870.1 | - | 10 - 11 | 1038 | 152 | |
CR749722.1 | - | 10 - 11 | 2023 | 152 | |
M23379.1 | - | 10 - 11 | 1577 | 152 | |
M23612.1 | - | 10 - 11 | 1027 | 152 |