The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002890.2 | NP_002881.1 | 17 - 18 | 2547 | 128 |
NM_022650.2 | NP_072179.1 | 17 - 18 | 1941 | 128 | |
XM_011543525.1 | XP_011541827.1 | 17 - 18 | 2999 | 128 | |
GenBank mRNA | AB209106.1 | - | 17 - 18 | 2547 | 128 |
AK300263.1 | - | 18 - 19 | 2048 | 128 | |
AK300269.1 | - | 17 - 18 | 1874 | 128 | |
AK300406.1 | - | 18 - 19 | 1967 | 128 | |
AK312739.1 | - | 17 - 18 | 1926 | 128 | |
BC033015.2 | - | 17 - 18 | 2535 | 128 | |
BC107870.1 | - | 17 - 18 | 1927 | 128 | |
CR749722.1 | - | 18 - 19 | 3057 | 128 | |
M23379.1 | - | 17 - 18 | 2466 | 128 | |
M23612.1 | - | 17 - 18 | 1916 | 128 |