The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002890.2 | NP_002881.1 | 19 - 20 | 2803 | 94 |
NM_022650.2 | NP_072179.1 | 19 - 20 | 2197 | 94 | |
GenBank mRNA | AB209106.1 | - | 19 - 20 | 2804 | 94 |
AK300263.1 | - | 20 - 21 | 2304 | 94 | |
AK300269.1 | - | 19 - 20 | 2130 | 94 | |
AK300406.1 | - | 20 - 21 | 2223 | 94 | |
AK312739.1 | - | 19 - 20 | 2182 | 94 | |
BC033015.2 | - | 19 - 20 | 2791 | 94 | |
BC107870.1 | - | 19 - 20 | 2183 | 94 | |
CR749722.1 | - | 20 - 21 | 3313 | 94 | |
M23379.1 | - | 19 - 20 | 2722 | 94 | |
M23612.1 | - | 19 - 20 | 2172 | 94 |