Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASA1
Entrez Gene ID
5921
Gene Name
RAS p21 protein activator 1
Gene Aliases
CM-AVM, CMAVM, GAP, PKWS, RASA, RASGAP, p120, p120GAP, p120RASGAP
Chromosome Location
Chr.5: 87267801 - 87391926 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002890.2 NP_002881.1 21 - 22 2953 73
NM_022650.2 NP_072179.1 21 - 22 2347 73
XM_011543525.1 XP_011541827.1 20 - 21 3318 73
GenBank mRNA AB209106.1 - 21 - 22 2954 73
AK300263.1 - 22 - 23 2454 73
AK300269.1 - 21 - 22 2280 73
AK300406.1 - 22 - 23 2373 73
AK312739.1 - 21 - 22 2332 73
BC033015.2 - 21 - 22 2941 73
BC107870.1 - 21 - 22 2333 73
CR749722.1 - 22 - 23 3463 73
M23379.1 - 21 - 22 2872 73
M23612.1 - 21 - 22 2322 73

View Full Product Details