The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002890.2 | NP_002881.1 | 8 - 9 | 1451 | 128 |
NM_022650.2 | NP_072179.1 | 8 - 9 | 845 | 128 | |
XM_011543525.1 | XP_011541827.1 | 8 - 9 | 1903 | 128 | |
XM_011543527.2 | XP_011541829.1 | 8 - 9 | 1903 | 128 | |
GenBank mRNA | AB209106.1 | - | 8 - 9 | 1451 | 128 |
AK300263.1 | - | 9 - 10 | 952 | 128 | |
AK300269.1 | - | 8 - 9 | 778 | 128 | |
AK300406.1 | - | 9 - 10 | 871 | 128 | |
AK312739.1 | - | 8 - 9 | 830 | 128 | |
BC020761.1 | - | 8 - 9 | 832 | 128 | |
BC033015.2 | - | 8 - 9 | 1439 | 128 | |
BC054891.1 | - | 8 - 9 | 811 | 128 | |
BC107870.1 | - | 8 - 9 | 831 | 128 | |
CR749722.1 | - | 8 - 9 | 1816 | 128 | |
M23379.1 | - | 8 - 9 | 1370 | 128 | |
M23612.1 | - | 8 - 9 | 820 | 128 |