Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TFDP2
Entrez Gene ID
7029
Gene Name
transcription factor Dp-2
Gene Aliases
DP2
Chromosome Location
Chr.3: 141944428 - 142149544 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178138.1 NP_001171609.1 11 - 12 1376 120
NM_001178139.1 NP_001171610.1 10 - 11 1338 120
NM_001178140.1 NP_001171611.1 7 - 8 834 120
NM_001178141.1 NP_001171612.1 7 - 8 767 120
NM_001178142.1 NP_001171613.1 5 - 6 615 120
NM_006286.4 NP_006277.1 8 - 9 892 120
XM_005247731.2 XP_005247788.1 10 - 11 1341 120
XM_011513101.2 XP_011511403.1 11 - 12 1437 120
XM_011513102.2 XP_011511404.1 9 - 10 1010 120
XM_011513106.2 XP_011511408.1 11 - 12 1434 120
XM_011513107.1 XP_011511409.1 10 - 11 1341 120
XM_011513108.2 XP_011511410.1 8 - 9 1164 120
XM_011513109.2 XP_011511411.1 6 - 7 736 120
XM_011513111.2 XP_011511413.1 6 - 7 733 120
XM_017007091.1 XP_016862580.1 10 - 11 1349 120
XM_017007092.1 XP_016862581.1 13 - 14 1595 120
XM_017007093.1 XP_016862582.1 13 - 14 1784 120
XM_017007094.1 XP_016862583.1 10 - 11 1370 120
XM_017007095.1 XP_016862584.1 12 - 13 1560 120
XM_017007096.1 XP_016862585.1 9 - 10 1267 120
XM_017007097.1 XP_016862586.1 11 - 12 1366 120
XM_017007098.1 XP_016862587.1 11 - 12 1514 120
XM_017007099.1 XP_016862588.1 12 - 13 1489 120
XM_017007100.1 XP_016862589.1 10 - 11 1338 120
XM_017007101.1 XP_016862590.1 10 - 11 1322 120
XM_017007102.1 XP_016862591.1 8 - 9 1117 120
XM_017007104.1 XP_016862593.1 8 - 9 1164 120
XM_017007105.1 XP_016862594.1 7 - 8 1094 120
GenBank mRNA AK303181.1 - 5 - 6 576 120
AK303634.1 - 7 - 8 709 120
AL080206.1 - 7 - 8 794 120
BC021113.1 - 7 - 8 796 120
L40386.1 - 7 - 8 733 120
U18422.1 - 8 - 9 844 120

View Full Product Details