Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBE3A
Entrez Gene ID
7337
Gene Name
ubiquitin protein ligase E3A
Gene Aliases
ANCR, AS, E6-AP, EPVE6AP, HPVE6A
Chromosome Location
Chr.15: 25337234 - 25439086 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000462.3 NP_000453.2 11 - 12 3047 137
NM_130838.1 NP_570853.1 7 - 8 2262 137
NM_130839.2 NP_570854.1 10 - 11 2982 137
XM_005268267.4 XP_005268324.1 12 - 13 3349 137
XM_005268268.4 XP_005268325.1 11 - 12 3236 137
XM_005268269.4 XP_005268326.1 12 - 13 3080 137
XM_005268270.4 XP_005268327.1 11 - 12 3037 137
XM_005268271.4 XP_005268328.1 8 - 9 2856 137
XM_006720675.3 XP_006720738.1 12 - 13 3301 137
XM_006720676.3 XP_006720739.1 9 - 10 2546 137
XM_011521994.2 XP_011520296.1 12 - 13 3238 137
XM_011521995.2 XP_011520297.1 12 - 13 2912 137
XM_017022544.1 XP_016878033.1 13 - 14 3274 137
XM_017022545.1 XP_016878034.1 12 - 13 3233 137
XM_017022546.1 XP_016878035.1 12 - 13 3141 137
XM_017022547.1 XP_016878036.1 11 - 12 3173 137
XM_017022548.1 XP_016878037.1 11 - 12 3168 137
XM_017022549.1 XP_016878038.1 11 - 12 2975 137
XM_017022550.1 XP_016878039.1 11 - 12 3127 137
XM_017022551.1 XP_016878040.1 12 - 13 3178 137
XM_017022552.1 XP_016878041.1 12 - 13 2868 137
XM_017022553.1 XP_016878042.1 12 - 13 3183 137
XM_017022554.1 XP_016878043.1 8 - 9 3187 137
XM_017022555.1 XP_016878044.1 10 - 11 2956 137
GenBank mRNA AB194232.1 - 9 - 10 2358 137
AF273050.1 - 8 - 9 2317 137
AK130491.1 - 6 - 7 1991 137
AK291405.1 - 8 - 9 2408 137
AK292514.1 - 10 - 11 2592 137
BC002582.2 - 10 - 11 137
BC009271.1 - 5 - 6 1539 137
U84404.1 - 12 - 13 2804 137
X98031.1 - 8 - 9 2287 137
X98032.1 - 6 - 7 2218 137
X98033.1 - 8 - 9 2278 137

View Full Product Details