The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242902.1 | NP_001229831.1 | 5 - 6 | 2255 | 61 |
NM_001242903.1 | NP_001229832.1 | 5 - 6 | 2528 | 61 | |
NM_003488.3 | NP_003479.1 | 4 - 5 | 2204 | 61 | |
XM_005257707.2 | XP_005257764.1 | 5 - 6 | 2524 | 61 | |
XM_005257709.2 | XP_005257766.1 | 4 - 5 | 2316 | 61 | |
XM_017025189.1 | XP_016880678.1 | 5 - 6 | 2262 | 61 | |
XM_017025190.1 | XP_016880679.1 | 4 - 5 | 2425 | 61 | |
XM_017025191.1 | XP_016880680.1 | 4 - 5 | 2061 | 61 | |
GenBank mRNA | AK292416.1 | - | 4 - 5 | 2183 | 61 |
AK297810.1 | - | 5 - 6 | 2254 | 61 | |
AK308397.1 | - | 5 - 6 | 2337 | 61 | |
BC000729.2 | - | 4 - 5 | 2136 | 61 | |
U34074.1 | - | 5 - 6 | 2138 | 61 | |
X97335.1 | - | 4 - 5 | 2097 | 61 |