The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001041.3 | NP_001032.2 | 35 - 36 | 4248 | 153 |
XM_011513078.2 | XP_011511380.1 | 35 - 36 | 4252 | 153 | |
GenBank mRNA | BC115034.1 | - | 35 - 36 | 4261 | 153 |
BC116452.1 | - | 35 - 36 | 4261 | 153 | |
BC116453.1 | - | 35 - 36 | 4261 | 153 | |
BC132834.1 | - | 35 - 36 | 4241 | 153 | |
BC132860.1 | - | 35 - 36 | 4241 | 153 | |
X63597.1 | - | 35 - 36 | 4247 | 153 |