The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001041.3 | NP_001032.2 | 40 - 41 | 4755 | 160 |
XM_011513078.2 | XP_011511380.1 | 40 - 41 | 4759 | 160 | |
GenBank mRNA | BC115034.1 | - | 40 - 41 | 4768 | 160 |
BC116452.1 | - | 40 - 41 | 4768 | 160 | |
BC116453.1 | - | 40 - 41 | 4768 | 160 | |
BC132834.1 | - | 40 - 41 | 4748 | 160 | |
BC132860.1 | - | 40 - 41 | 4748 | 160 | |
X63597.1 | - | 40 - 41 | 4754 | 160 |