The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320227.1 | NP_001307156.1 | 10 - 11 | 1016 | 75 |
NM_015938.4 | NP_057022.2 | 10 - 11 | 1016 | 75 | |
XM_005247511.1 | XP_005247568.1 | 10 - 11 | 966 | 75 | |
XM_005247512.1 | XP_005247569.1 | 10 - 11 | 1297 | 75 | |
GenBank mRNA | AF132941.1 | - | 10 - 11 | 987 | 75 |
AK021775.1 | - | 7 - 8 | 627 | 75 | |
AK094749.1 | - | 9 - 10 | 1319 | 75 | |
AK225664.1 | - | 10 - 11 | 998 | 75 | |
AK296710.1 | - | 9 - 10 | 624 | 75 | |
AK308865.1 | - | 9 - 10 | 797 | 75 | |
BC013317.1 | - | 10 - 11 | 999 | 75 |