The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243146.1 | NP_001230075.1 | 8 - 9 | 802 | 137 |
NM_001243147.1 | NP_001230076.1 | 9 - 10 | 853 | 137 | |
NM_002533.3 | NP_002524.2 | 10 - 11 | 1126 | 137 | |
NM_206840.2 | NP_996671.1 | 9 - 10 | 1052 | 137 | |
XM_011544196.1 | XP_011542498.1 | 11 - 12 | 1190 | 137 | |
XM_011544198.1 | XP_011542500.1 | 10 - 11 | 1145 | 137 | |
XM_011544199.1 | XP_011542501.1 | 10 - 11 | 1099 | 137 | |
XM_011544200.1 | XP_011542502.1 | 9 - 10 | 1073 | 137 | |
XM_017001378.1 | XP_016856867.1 | 12 - 13 | 1299 | 137 | |
XM_017001379.1 | XP_016856868.1 | 11 - 12 | 1218 | 137 | |
XM_017001380.1 | XP_016856869.1 | 12 - 13 | 1299 | 137 | |
XM_017001381.1 | XP_016856870.1 | 12 - 13 | 1296 | 137 | |
XM_017001382.1 | XP_016856871.1 | 11 - 12 | 1229 | 137 | |
XM_017001383.1 | XP_016856872.1 | 10 - 11 | 1148 | 137 | |
XM_017001384.1 | XP_016856873.1 | 12 - 13 | 1299 | 137 | |
XM_017001385.1 | XP_016856874.1 | 12 - 13 | 1299 | 137 | |
XM_017001386.1 | XP_016856875.1 | 8 - 9 | 992 | 137 | |
GenBank mRNA | AK297070.1 | - | 10 - 11 | 1085 | 137 |
AK297396.1 | - | 8 - 9 | 753 | 137 | |
AK298244.1 | - | 9 - 10 | 802 | 137 | |
AK298826.1 | - | 9 - 10 | 1004 | 137 | |
BC012105.1 | - | 8 - 9 | 713 | 137 | |
U68140.1 | - | 10 - 11 | 1120 | 137 | |
U78772.1 | - | 9 - 10 | 1033 | 137 |