The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243146.1 | NP_001230075.1 | 18 - 19 | 2107 | 71 |
NM_001243147.1 | NP_001230076.1 | 19 - 20 | 2158 | 71 | |
NM_002533.3 | NP_002524.2 | 20 - 21 | 2431 | 71 | |
NM_206840.2 | NP_996671.1 | 19 - 20 | 2357 | 71 | |
XM_011544196.1 | XP_011542498.1 | 21 - 22 | 2495 | 71 | |
XM_011544198.1 | XP_011542500.1 | 20 - 21 | 2450 | 71 | |
XM_011544199.1 | XP_011542501.1 | 20 - 21 | 2404 | 71 | |
XM_011544200.1 | XP_011542502.1 | 19 - 20 | 2378 | 71 | |
XM_017001378.1 | XP_016856867.1 | 22 - 23 | 2604 | 71 | |
XM_017001379.1 | XP_016856868.1 | 21 - 22 | 2523 | 71 | |
XM_017001380.1 | XP_016856869.1 | 22 - 23 | 2604 | 71 | |
XM_017001381.1 | XP_016856870.1 | 22 - 23 | 2601 | 71 | |
XM_017001382.1 | XP_016856871.1 | 21 - 22 | 2534 | 71 | |
XM_017001383.1 | XP_016856872.1 | 20 - 21 | 2453 | 71 | |
XM_017001386.1 | XP_016856875.1 | 18 - 19 | 2297 | 71 | |
GenBank mRNA | AK297070.1 | - | 20 - 21 | 2390 | 71 |
AK297396.1 | - | 18 - 19 | 2058 | 71 | |
AK298244.1 | - | 19 - 20 | 2107 | 71 | |
AK298826.1 | - | 18 - 19 | 2209 | 71 | |
BC012105.1 | - | 18 - 19 | 2018 | 71 | |
BU683548.1 | - | 3 - 4 | 71 | ||
U68140.1 | - | 20 - 21 | 2425 | 71 | |
U78772.1 | - | 19 - 20 | 2338 | 71 |