Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL14A1
Entrez Gene ID
7373
Gene Name
collagen type XIV alpha 1 chain
Gene Aliases
UND
Chromosome Location
Chr.8: 120124468 - 120373575 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021110.3 NP_066933.1 47 - 48 5580 68
XM_006716651.3 XP_006716714.1 47 - 48 5443 68
XM_017013809.1 XP_016869298.1 47 - 48 5369 68
GenBank mRNA BC014640.1 - 26 - 27 2579 68
BC083495.1 - 13 - 14 1086 68
BC140893.1 - 46 - 47 5349 68

View Full Product Details