The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021110.3 | NP_066933.1 | 26 - 27 | 3482 | 66 |
XM_005251059.3 | XP_005251116.1 | 26 - 27 | 3482 | 66 | |
XM_006716651.3 | XP_006716714.1 | 26 - 27 | 3345 | 66 | |
XM_017013809.1 | XP_016869298.1 | 26 - 27 | 3271 | 66 | |
GenBank mRNA | AK293080.1 | - | 26 - 27 | 3452 | 66 |
AK307716.1 | - | 26 - 27 | 3446 | 66 | |
BC014640.1 | - | 5 - 6 | 481 | 66 | |
BC140893.1 | - | 25 - 26 | 3251 | 66 | |
Y11710.1 | - | 1 - 2 | 139 | 66 |