Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL14A1
Entrez Gene ID
7373
Gene Name
collagen type XIV alpha 1 chain
Gene Aliases
UND
Chromosome Location
Chr.8: 120124468 - 120373575 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021110.3 NP_066933.1 29 - 30 3914 104
XM_005251059.3 XP_005251116.1 29 - 30 3914 104
XM_006716651.3 XP_006716714.1 29 - 30 3777 104
XM_017013809.1 XP_016869298.1 29 - 30 3703 104
GenBank mRNA BC014640.1 - 8 - 9 913 104
BC140893.1 - 28 - 29 3683 104
Y11710.1 - 4 - 5 571 104

View Full Product Details