Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL14A1
Entrez Gene ID
7373
Gene Name
collagen type XIV alpha 1 chain
Gene Aliases
UND
Chromosome Location
Chr.8: 120124468 - 120373575 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021110.3 NP_066933.1 6 - 7 867 113
XM_005251059.3 XP_005251116.1 6 - 7 867 113
XM_006716651.3 XP_006716714.1 6 - 7 730 113
XM_017013809.1 XP_016869298.1 6 - 7 656 113
GenBank mRNA AK293080.1 - 6 - 7 837 113
AK307716.1 - 6 - 7 831 113
AK311498.1 - 6 - 7 831 113
BC036192.1 - 9 - 10 893 113
BC140893.1 - 5 - 6 636 113
BQ717750.1 - 4 - 5 520 113
M64108.1 - 1 - 2 36 113

View Full Product Details