The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143964.3 | NP_001137436.1 | 7 - 8 | 1068 | 74 |
NM_001143965.3 | NP_001137437.1 | 7 - 8 | 928 | 74 | |
NM_001143966.3 | NP_001137438.1 | 6 - 7 | 847 | 74 | |
NM_001258457.2 | NP_001245386.1 | 6 - 7 | 790 | 74 | |
NM_001318805.1 | NP_001305734.1 | 7 - 8 | 985 | 74 | |
NM_016495.5 | NP_057579.1 | 7 - 8 | 1088 | 74 | |
GenBank mRNA | AB449888.1 | - | 6 - 7 | 792 | 74 |
AF151073.1 | - | 7 - 8 | 885 | 74 | |
AK223445.1 | - | 7 - 8 | 965 | 74 | |
AY542308.1 | - | 6 - 7 | 712 | 74 | |
AY826820.1 | - | 6 - 7 | 773 | 74 | |
BC007054.1 | - | 7 - 8 | 889 | 74 | |
BC050465.1 | - | 6 - 7 | 789 | 74 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318809.1 | NP_001305738.1 | 7 - 8 | 928 | 74 |
GenBank mRNA | AK057228.1 | - | 7 - 8 | 888 | 74 |
BU620048.1 | - | 2 - 3 | 74 |