The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320759.1 | NP_001307688.1 | 4 - 5 | 742 | 66 |
NM_004618.4 | NP_004609.1 | 5 - 6 | 816 | 66 | |
XM_011524001.2 | XP_011522303.1 | 1 - 2 | 115 | 66 | |
GenBank mRNA | AK126869.1 | - | 2 - 3 | 381 | 66 |
AK292926.1 | - | 5 - 6 | 639 | 66 | |
AK296437.1 | - | 4 - 5 | 574 | 66 | |
AK311668.1 | - | 3 - 4 | 583 | 66 | |
BC051748.1 | - | 5 - 6 | 698 | 66 | |
U43431.1 | - | 5 - 6 | 679 | 66 |