Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSNK1G3
Entrez Gene ID
1456
Gene Name
casein kinase 1 gamma 3
Gene Aliases
CKI-gamma 3, CSNK1G3L
Chromosome Location
Chr.5: 123512099 - 123617045 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031812.3 NP_001026982.1 12 - 13 1938 77
NM_001044723.2 NP_001038188.1 12 - 13 1562 77
NM_001270572.1 NP_001257501.1 11 - 12 1469 77
NM_001270573.1 NP_001257502.1 11 - 12 1513 77
NM_001270574.1 NP_001257503.1 9 - 10 1405 77
XM_005271891.1 XP_005271948.1 13 - 14 2037 77
XM_005271892.2 XP_005271949.1 13 - 14 2034 77
XM_005271895.2 XP_005271952.1 12 - 13 1941 77
XM_017009057.1 XP_016864546.1 14 - 15 2112 77
XM_017009058.1 XP_016864547.1 16 - 17 2321 77
XM_017009059.1 XP_016864548.1 15 - 16 2180 77
XM_017009060.1 XP_016864549.1 15 - 16 2236 77
XM_017009061.1 XP_016864550.1 14 - 15 2109 77
XM_017009062.1 XP_016864551.1 14 - 15 1668 77
XM_017009064.1 XP_016864553.1 13 - 14 2016 77
XM_017009065.1 XP_016864554.1 13 - 14 2013 77
XM_017009066.1 XP_016864555.1 14 - 15 2081 77
XM_017009069.1 XP_016864558.1 13 - 14 2009 77
GenBank mRNA AF049090.1 - 12 - 13 77
AK289405.1 - 11 - 12 1414 77
AK299739.1 - 11 - 12 1352 77
AK316019.1 - 9 - 10 1244 77
BC047567.1 - 12 - 13 1590 77

View Full Product Details