The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002292.3 | NP_002283.3 | 13 - 14 | 2030 | 143 |
XM_005265127.3 | XP_005265184.1 | 14 - 15 | 1829 | 143 | |
GenBank mRNA | S77512.1 | - | 13 - 14 | 1883 | 143 |
X79683.1 | - | 14 - 15 | 1896 | 143 |