The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002292.3 | NP_002283.3 | 5 - 6 | 946 | 108 |
XM_005265127.3 | XP_005265184.1 | 6 - 7 | 745 | 108 | |
GenBank mRNA | S77512.1 | - | 5 - 6 | 799 | 108 |
X79683.1 | - | 6 - 7 | 812 | 108 |