The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002292.3 | NP_002283.3 | 7 - 8 | 1218 | 59 |
XM_005265127.3 | XP_005265184.1 | 8 - 9 | 1017 | 59 | |
GenBank mRNA | S77512.1 | - | 7 - 8 | 1071 | 59 |
X79683.1 | - | 8 - 9 | 1084 | 59 |