Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EHMT1
Entrez Gene ID
79813
Gene Name
euchromatic histone lysine methyltransferase 1
Gene Aliases
EUHMTASE1, Eu-HMTase1, FP13812, GLP, GLP1, KMT1D, bA188C12.1
Chromosome Location
Chr.9: 137618988 - 137836127 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145527.1 NP_001138999.1 13 - 14 2234 89
NM_024757.4 NP_079033.4 13 - 14 2234 89
XM_005266105.4 XP_005266162.1 13 - 14 2277 89
XM_005266110.1 XP_005266167.1 12 - 13 2149 89
XM_006717288.2 XP_006717351.1 13 - 14 2195 89
XM_011519021.2 XP_011517323.1 13 - 14 3232 89
XM_011519022.2 XP_011517324.1 13 - 14 2687 89
XM_011519023.2 XP_011517325.1 13 - 14 3211 89
XM_011519024.2 XP_011517326.1 12 - 13 3155 89
XM_011519025.1 XP_011517327.1 13 - 14 2397 89
XM_011519026.2 XP_011517328.1 12 - 13 3090 89
XM_011519027.2 XP_011517329.1 13 - 14 3233 89
XM_011519028.2 XP_011517330.1 13 - 14 3233 89
XM_011519029.2 XP_011517331.1 5 - 6 822 89
XM_011519033.2 XP_011517335.1 12 - 13 3069 89
XM_017015134.1 XP_016870623.1 13 - 14 2665 89
XM_017015135.1 XP_016870624.1 13 - 14 2217 89
XM_017015136.1 XP_016870625.1 12 - 13 2139 89
XM_017015137.1 XP_016870626.1 13 - 14 2376 89
XM_017015138.1 XP_016870627.1 12 - 13 2128 89
GenBank mRNA AB028932.1 - 8 - 9 1054 89
AB058779.2 - 8 - 9 1054 89
AK022941.1 - 12 - 13 2149 89
AY083210.1 - 11 - 12 2104 89
BC011608.2 - 3 - 4 451 89
BC047504.1 - 13 - 14 2234 89
BM681619.1 - 3 - 4 89

View Full Product Details