Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EHMT1
Entrez Gene ID
79813
Gene Name
euchromatic histone lysine methyltransferase 1
Gene Aliases
EUHMTASE1, Eu-HMTase1, FP13812, GLP, GLP1, KMT1D, bA188C12.1
Chromosome Location
Chr.9: 137618988 - 137836127 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024757.4 NP_079033.4 20 - 21 3072 104
XM_005266105.4 XP_005266162.1 20 - 21 3115 104
XM_005266110.1 XP_005266167.1 19 - 20 2987 104
XM_006717288.2 XP_006717351.1 20 - 21 3033 104
XM_011519021.2 XP_011517323.1 20 - 21 4070 104
XM_011519022.2 XP_011517324.1 20 - 21 3525 104
XM_011519023.2 XP_011517325.1 20 - 21 4049 104
XM_011519024.2 XP_011517326.1 19 - 20 3993 104
XM_011519025.1 XP_011517327.1 20 - 21 3235 104
XM_011519026.2 XP_011517328.1 19 - 20 3928 104
XM_011519029.2 XP_011517331.1 12 - 13 1660 104
XM_011519030.2 XP_011517332.1 6 - 7 1355 104
XM_011519031.1 XP_011517333.1 6 - 7 713 104
XM_011519033.2 XP_011517335.1 19 - 20 3907 104
XM_017015134.1 XP_016870623.1 20 - 21 3503 104
XM_017015135.1 XP_016870624.1 20 - 21 3055 104
XM_017015136.1 XP_016870625.1 19 - 20 2977 104
XM_017015137.1 XP_016870626.1 20 - 21 3214 104
XM_017015138.1 XP_016870627.1 19 - 20 2966 104
GenBank mRNA AB028932.1 - 15 - 16 1892 104
AB058779.2 - 15 - 16 1892 104
AY083210.1 - 18 - 19 2942 104
BC011608.2 - 10 - 11 1289 104

View Full Product Details