The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165903.1 | NP_001159375.1 | 1 - 2 | 83 | 77 |
NM_004603.3 | NP_004594.1 | 1 - 2 | 83 | 77 | |
XM_017012567.1 | XP_016868056.1 | 1 - 2 | 98 | 77 | |
GenBank mRNA | AK299959.1 | - | 1 - 2 | 57 | 77 |
BC003011.1 | - | 1 - 2 | 55 | 77 | |
BC064644.1 | - | 1 - 2 | 54 | 77 | |
DA159446.1 | - | 1 - 2 | 83 | 77 |